4(1H)-Pyrimidinone ≥98%

Supplier: Ambeed
Danger

Synonyms: 3,4-Dihydropyrimidin-4-one, 1H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(1H)-one

A457375-25G A457375-5G
77748-925EA 229 USD
77748-925 77748-926
4(1H)-Pyrimidinone ≥98%
4(1H)-Pyrimidinone

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.09 g/mol
MDL Number: MFCD00006664
CAS Number: 51953-17-4
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR