(Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R,E)-3-hydroxy-4-(3-(trifluoromethyl)phenoxy)but-1-en-1-yl)cyclopentyl)hept-5-enoate ≥98%

Supplier: Ambeed
Warning

Synonyms: Travoprost

A465015-250MG A465015-100MG
77602-032EA 273.68 USD
77602-032 77669-088
(Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R,E)-3-hydroxy-4-(3-(trifluoromethyl)phenoxy)but-1-en-1-yl)cyclopentyl)hept-5-enoate ≥98%
(Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R,E)-3-hydroxy-4-(3-(trifluoromethyl)phenoxy)but-1-en-1-yl)cyclopentyl)hept-5-enoate

Precaución: for Research Use Only

Formula: C₂₆H₃₅F₃O₆
MW: 500.55 g/mol
MDL Number: MFCD03411995
CAS Number: 157283-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR