1,4-Naphthoquinone ≥97%

Supplier: Ambeed
Danger

Synonyms: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene

A457360-100G A457360-25G
77594-438EA 169.13 USD
77594-438 77594-440
1,4-Naphthoquinone ≥97%
1,4-Naphthoquinone

Precaución: for Research Use Only

Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Boiling Pt: 298
Melting Pt: 124
Flash Pt: 141
MDL Number: MFCD00001676
CAS Number: 130-15-4
UN: 2923
ADR: 8,6.1 Ⅲ
Merck Index: 12,06482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR