N-Hexanoyl-D-erythro-sphingosine ≥97%

Supplier: Ambeed
Warning

Synonyms: C6 ceramide, Caproyl ceramide, N-((2S,3R,E)-1,3-Dihydroxyoctadec-4-en-2-yl)hexanamide

A464869-10MG A464869-1MG A464869-5MG
77602-026EA 112.75 USD
77602-026 77602-028 77602-030
N-Hexanoyl-D-erythro-sphingosine ≥97%
N-Hexanoyl-D-erythro-sphingosine

Precaución: for Research Use Only

Formula: C₂₄H₄₇NO₃
MW: 397.644 g/mol
MDL Number: MFCD00870174
CAS Number: 124753-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR