4-Bromo-2,6-dimethylphenylboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-2,6-dimethylphenyl)boronic acid

A456961-100MG A456961-250MG
77673-154EA 33 USD
77673-154 77673-156
4-Bromo-2,6-dimethylphenylboronic acid ≥95%
4-Bromo-2,6-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₂
MW: 228.879 g/mol
MDL Number: MFCD13181651
CAS Number: 1160561-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR