(1S,1'S)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene ≥98%

Supplier: Ambeed
Warning

Synonyms: (1Rp,1'Rp)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene

A456941-100MG A456941-250MG
77594-436EA 181.43 USD
77594-436 77616-972
(1S,1'S)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene ≥98%
(1S,1'S)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene

Precaución: for Research Use Only

Formula: C₆₄H₇₄FeN₂O₄P₂
MW: 1053.08 g/mol
MDL Number: MFCD08561145
CAS Number: 849925-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR