3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

A456828-1G A456828-250MG A456828-5G
77886-198EA 174 USD
77886-198 77886-199 77886-200
3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid ≥98%
3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₄
MDL Number: MFCD08436059
CAS Number: 913835-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR