Ethyl (2-amino-6-((4-fluorobenzyl)amino)pyridin-3-yl)carbamate ≥98%

Supplier: Ambeed
Warning

Synonyms: Flupirtine

A456752-10MG A456752-1MG A456752-50MG
77601-974EA 37.93 USD
77601-974 77601-976 77626-906
Ethyl (2-amino-6-((4-fluorobenzyl)amino)pyridin-3-yl)carbamate ≥98%
Ethyl (2-amino-6-((4-fluorobenzyl)amino)pyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₇FN₄O₂
MW: 304.32 g/mol
MDL Number: MFCD00864265
CAS Number: 56995-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR