H-Glu-Otbu·HCl ≥98%

Supplier: Ambeed
Warning

A456556-5G A456556-25G A456556-1G
77601-972EA 86.1 USD
77601-972 77669-054 77669-052
H-Glu-Otbu·HCl ≥98%
H-Glu-Otbu·HCl

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₄
MDL Number: MFCD02684516
CAS Number: 144313-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR