Methyl 6-fluoro-5-methylpyridine-3-carboxylate ≥98%

Supplier: Ambeed
Danger

A463823-250MG A463823-5G
77890-940EA 45 USD
77890-940 77890-941
Methyl 6-fluoro-5-methylpyridine-3-carboxylate ≥98%
Methyl 6-fluoro-5-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD11616889
CAS Number: 211122-38-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR