1-Phenyl-3,3-bis(pyridin-4-ylmethyl)indolin-2-one dihydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Linopirdine dihydrochloride
A463680-25G
A463680-5G
A463680-1G
A463680-250MG
77617-006EA
2260.85
USD
77617-006
77617-008
77746-949
77746-950
1-Phenyl-3,3-bis(pyridin-4-ylmethyl)indolin-2-one dihydrochloride ≥98%
1-Phenyl-3,3-bis(pyridin-4-ylmethyl)indolin-2-one dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₆H₂₃Cl₂N₃O MW: 464.38 g/mol |
MDL Number:
MFCD06245587 CAS Number: 113168-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...