O-(tert-Butyldimethylsilyl)hydroxylamine ≥98%

Supplier: Ambeed
Danger

A463419-1G A463419-5G
77602-016EA 76.88 USD
77602-016 77669-086
O-(tert-Butyldimethylsilyl)hydroxylamine ≥98%
O-(tert-Butyldimethylsilyl)hydroxylamine

Precaución: for Research Use Only

Formula: C₆H₁₇NOSi
MW: 147.293 g/mol
MDL Number: MFCD00054944
CAS Number: 41879-39-4
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR