KN-62 ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-4-(2-(N-Methylisoquinoline-5-sulphonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl)phenyl isoquinoline-5-sulphonate

A455366-10MG A455366-1MG A455366-5MG
77601-962EA 69.7 USD
77601-962 77601-964 77601-966
KN-62 ≥98%
KN-62

Precaución: for Research Use Only

Formula: C₃₈H₃₅N₅O₆S₂
MW: 721.84 g/mol
MDL Number: MFCD00083180
CAS Number: 127191-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR