6-Bromo-2-fluoro-3-nitropyridine ≥95%

Supplier: Ambeed
Warning

A462821-1G A462821-5G A462821-250MG
77617-004EA 122.57 USD
77617-004 77748-944 77748-943
6-Bromo-2-fluoro-3-nitropyridine ≥95%
6-Bromo-2-fluoro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFN₂O₂
MDL Number: MFCD28669853
CAS Number: 1417333-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR