4-(7-(4-((Benzyloxy)carbonyl)piperazin-1-yl)-4,4-dimethyl-8-nitrochromeno[4,3-c]pyrazol-1(4H)-yl)benzoic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: Lin28-let-7a antagonist 1

A462770-10MG A462770-1MG A462770-5MG
77598-490EA 188.6 USD
77598-490 77598-492 77598-494
4-(7-(4-((Benzyloxy)carbonyl)piperazin-1-yl)-4,4-dimethyl-8-nitrochromeno[4,3-c]pyrazol-1(4H)-yl)benzoic acid ≥99%
4-(7-(4-((Benzyloxy)carbonyl)piperazin-1-yl)-4,4-dimethyl-8-nitrochromeno[4,3-c]pyrazol-1(4H)-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₃₁H₂₉N₅O₇
MW: 583.5913 g/mol
MDL Number: MFCD30741839
CAS Number: 2024548-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR