(S)-N2-(1-(4-Fluorophenyl)ethyl)-4-(1-methyl-1H-pyrazol-4-yl)-N6-(pyrazin-2-yl)pyridine-2,6-diamine maleate ≥98%

Supplier: Ambeed
Warning

Synonyms: Ilginatinib maleate

A454719-10MG A454719-1MG A454719-5MG
77616-960EA 96.82 USD
77616-960 77616-962 77616-964
(S)-N2-(1-(4-Fluorophenyl)ethyl)-4-(1-methyl-1H-pyrazol-4-yl)-N6-(pyrazin-2-yl)pyridine-2,6-diamine maleate ≥98%
(S)-N2-(1-(4-Fluorophenyl)ethyl)-4-(1-methyl-1H-pyrazol-4-yl)-N6-(pyrazin-2-yl)pyridine-2,6-diamine maleate

Precaución: for Research Use Only

Formula: C₂₅H₂₄FN₇O₄
MW: 505.5 g/mol
MDL Number: MFCD30343859
CAS Number: 1354799-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR