2-Fluoro-5-methoxypyridine-3-boronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-methoxypyridin-3-yl)boronic acid

A454654-1G A454654-5G
77884-169EA 289 USD
77884-169 77884-170
2-Fluoro-5-methoxypyridine-3-boronic acid 98+%
2-Fluoro-5-methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MW: 170.934 g/mol
MDL Number: MFCD16611243
CAS Number: 1253577-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR