(5-Chlorothiophen-2-yl)methanol ≥95%

Supplier: Ambeed
Warning

A462326-10G A462326-25G
77872-768EA 37 USD
77872-768 77872-769
(5-Chlorothiophen-2-yl)methanol ≥95%
(5-Chlorothiophen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅ClOS
MDL Number: MFCD06202649
CAS Number: 74168-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR