4-(Piperazin-1-ylmethyl)benzoic acid dihydrochloride ≥95%

Supplier: Ambeed
Warning

A454242-1G A454242-250MG A454242-5G
77890-925EA 92 USD
77890-925 77890-926 77890-927
4-(Piperazin-1-ylmethyl)benzoic acid dihydrochloride ≥95%
4-(Piperazin-1-ylmethyl)benzoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈Cl₂N₂O₂
MDL Number: MFCD09753579
CAS Number: 86620-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR