Ammonium 1-pyrrolidinecarbodithioate ≥98%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

A462174-500G A462174-100G A462174-100MG A462174-5G A462174-25G
77617-002EA 204.97 USD
77617-002 77746-941 77746-942 77746-944 77746-943
Ammonium 1-pyrrolidinecarbodithioate ≥98%
Ammonium 1-pyrrolidinecarbodithioate

Precaución: for Research Use Only

Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
MDL Number: MFCD00012720
CAS Number: 5108-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR