3-(2,4-Dihydroxy-3-(3-methylbut-2-en-1-yl)phenyl)-5,7-dihydroxy-4H-chromen-4-one ≥98%

Supplier: Ambeed
Warning

A462091-1MG A462091-5MG A462091-10MG
77686-238EA 108 USD
77686-238 77686-239 77892-939
3-(2,4-Dihydroxy-3-(3-methylbut-2-en-1-yl)phenyl)-5,7-dihydroxy-4H-chromen-4-one ≥98%
3-(2,4-Dihydroxy-3-(3-methylbut-2-en-1-yl)phenyl)-5,7-dihydroxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₆
MDL Number: MFCD04039831
CAS Number: 66056-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR