Erythromycin ethylsuccinate ≥765 IU/mg

Supplier: Ambeed
Danger

Synonyms: (2S,3R,4S,6R)-4-(Dimethylamino)-2-(((3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,7,9,11,13-hexamethyl-2,10-dioxooxacyclotetradecan-6-yl)oxy)-6-methyltet

A453784-1MG A453784-50MG A453784-250MG
77669-042EA 5 USD
77669-042 77669-044 77879-993
Erythromycin ethylsuccinate ≥765 IU/mg
Erythromycin ethylsuccinate

Precaución: for Research Use Only

Formula: C₄₃H₇₅NO₁₆
MDL Number: MFCD00084692
CAS Number: 1264-62-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR