Tributylphosphonium tetrafluoroborate ≥98%

Supplier: Ambeed
Warning

Synonyms: Tri-n-butylphosphonium tetrafluoroborate

A453607-1G A453607-250MG A453607-25G A453607-5G
77572-430EA 14.42 USD
77572-430 77601-952 77601-954 77601-956
Tributylphosphonium tetrafluoroborate ≥98%
Tributylphosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₂H₂₈BF₄P
MW: 290.13 g/mol
MDL Number: MFCD05664257
CAS Number: 113978-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR