N-(2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)phenyl)-2-fluoro-3-(N-methylbenzamido)benzamide ≥99%

Supplier: Ambeed
Danger

Synonyms: Broflanilide

A453417-1MG A453417-5MG
77616-956EA 36.05 USD
77616-956 77616-958
N-(2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)phenyl)-2-fluoro-3-(N-methylbenzamido)benzamide ≥99%
N-(2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)phenyl)-2-fluoro-3-(N-methylbenzamido)benzamide

Precaución: for Research Use Only

Formula: C₂₅H₁₄BrF₁₁N₂O₂
MW: 663.28 g/mol
MDL Number: MFCD31657367
CAS Number: 1207727-04-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR