3-Methylpiperidin-3-ol hydrochloride ≥98%

Supplier: Ambeed
Warning

A461590-25G A461590-1G
77594-454EA 402.83 USD
77594-454 77613-430
3-Methylpiperidin-3-ol hydrochloride ≥98%
3-Methylpiperidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD17171256
CAS Number: 955028-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR