DDR1-IN-1 ≥99%

Supplier: Ambeed
Warning

Synonyms: 4-((4-ethylpiperazin-1-yl)methyl)-N-(4-methyl-3-(2-oxoindolin-5-yloxy)phenyl)-3-(trifluoromethyl)benzamide, 4-((4-Ethylpiperazin-1-yl)methyl)-N-(4-methyl-3-((2-oxoindolin-5-yl)oxy)phenyl)-3-(trifluoromethyl)benzamide

A461561-1MG A461561-5MG A461561-10MG
77616-998EA 27.81 USD
77616-998 77746-940 77746-939
DDR1-IN-1 ≥99%
DDR1-IN-1

Precaución: for Research Use Only

Formula: C₃₀H₃₁F₃N₄O₃
MW: 552.5982 g/mol
MDL Number: MFCD28144507
CAS Number: 1449685-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR