Ethyl sorbate ≥95%

Supplier: Ambeed
Danger

Synonyms: Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate

A461496-25G A461496-100G A461496-10G A461496-500G
77602-002EA 14.35 USD
77602-002 77601-998 77602-000 77669-076
Ethyl sorbate ≥95%
Ethyl sorbate

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00009296
CAS Number: 2396-84-1
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR