Sodium 2-methyl-2-propanethiolate ≥98%

Supplier: Ambeed
Danger

A453384-5G A453384-1G
77594-424EA 47.15 USD
77594-424 77872-745
Sodium 2-methyl-2-propanethiolate ≥98%
Sodium 2-methyl-2-propanethiolate

Precaución: for Research Use Only

Formula: C₄H₉NaS
MDL Number: MFCD00043311
CAS Number: 29364-29-2
UN: 3263
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR