Methyl-5-(bromomethyl)thiophene-2-carboxylate ≥95%

Supplier: Ambeed
Danger

Synonyms: Methyl 5-(bromomethyl)thiophene-2-carboxylate

A461321-100MG A461321-250MG
77882-180EA 61 USD
77882-180 77882-181
Methyl-5-(bromomethyl)thiophene-2-carboxylate ≥95%
Methyl-5-(bromomethyl)thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.11 g/mol
MDL Number: MFCD09834150
CAS Number: 108499-32-7
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR