4-(1H-Imidazol-5-yl)pyridine ≥95%

Supplier: Ambeed
Warning

A453362-250MG A453362-100MG
77594-422EA 53.3 USD
77594-422 77884-165
4-(1H-Imidazol-5-yl)pyridine ≥95%
4-(1H-Imidazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MDL Number: MFCD08668577
CAS Number: 51746-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR