3-Bromo-5-cyanobenzaldehyde ≥98%

Supplier: Ambeed
Warning

A461149-1G A461149-250MG
77882-178EA 79 USD
77882-178 77882-179
3-Bromo-5-cyanobenzaldehyde ≥98%
3-Bromo-5-cyanobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrNO
MDL Number: MFCD09261040
CAS Number: 644982-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR