N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥98%

Supplier: Ambeed
Danger

A453138-5G A453138-1G A453138-25G
77572-426EA 17 USD
77572-426 77882-166 77882-167
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥98%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide

Precaución: for Research Use Only

Formula: C₆H₁₂F₃NOSi
MDL Number: MFCD00000411
CAS Number: 24589-78-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR