7-Chloro-4-quinolineboronic acid pinacol ester ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
A452726-1G
A452726-250MG
77684-239EA
99
USD
77684-239
77684-240
7-Chloro-4-quinolineboronic acid pinacol ester ≥98%
7-Chloro-4-quinolineboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₅H₁₇BClNO₂ MW: 289.56 g/mol |
MDL Number:
MFCD07783020 CAS Number: 871125-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...