Methyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate ≥97%

Supplier: Ambeed
Warning

A452703-100MG A452703-250MG
77616-950EA 53.56 USD
77616-950 77616-952
Methyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate ≥97%
Methyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆BrN₃O₂
MW: 256.0561 g/mol
MDL Number: MFCD12400860
CAS Number: 1159811-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR