5-Methoxypyridine-3-boronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

A460399-5G A460399-1G A460399-25G
77594-448EA 79.95 USD
77594-448 77613-424 77890-935
5-Methoxypyridine-3-boronic acid ≥95%
5-Methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR