(Z)-Isopropyl 7-((1R,2R,3R,5S)-2-((E)-3,3-difluoro-4-phenoxybut-1-en-1-yl)-3,5-dihydroxycyclopentyl)hept-5-enoate ≥97%

Supplier: Ambeed
Warning

Synonyms: Tafluprost

A451878-100MG A451878-250MG
77673-120EA 230 USD
77673-120 77673-122
(Z)-Isopropyl 7-((1R,2R,3R,5S)-2-((E)-3,3-difluoro-4-phenoxybut-1-en-1-yl)-3,5-dihydroxycyclopentyl)hept-5-enoate ≥97%
(Z)-Isopropyl 7-((1R,2R,3R,5S)-2-((E)-3,3-difluoro-4-phenoxybut-1-en-1-yl)-3,5-dihydroxycyclopentyl)hept-5-enoate

Precaución: for Research Use Only

Formula: C₂₅H₃₄F₂O₅
MW: 452.53 g/mol
MDL Number: MFCD08062150
CAS Number: 209860-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR