AF353 98+%

Supplier: Ambeed
Warning

Synonyms: 5-(5-Iodo-2-isopropyl-4-methoxyphenoxy)pyrimidine-2,4-diamine, Ro-4,5-(5-iodo-4-methoxy-2-propan-2-ylphenoxy)pyrimidine-2,4-diamine, AF-353

A451786-100MG A451786-50MG
77686-217EA 277 USD
77686-217 77686-218
AF353 98+%
AF353

Precaución: for Research Use Only

Formula: C₁₄H₁₇IN₄O₂
MW: 400.22 g/mol
MDL Number: MFCD18782750
CAS Number: 865305-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR