1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98% stabilised

Supplier: Ambeed
Warning

Synonyms: Diethylene glycol divinyl ether, 1,1'-[Oxybis(ethyleneoxy)]diethene, 1,1'-(Oxybis(ethyleneoxy))diethylene, 1,1'-(Oxybis(ethyleneoxy))diethene, Diethyleneglycoldivinylether, (2-(2-Ethenyloxyethoxy)ethoxy)ethene

Stabilization: Stabilised with potassium hydroxide 0.1%

A451734-500G A451734-100G A451734-25G
77879-989EA 96 USD
77879-989 77879-987 77879-988
1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98% stabilised
1,1'-[Oxybis(ethyleneoxy)]diethylene

Precaución: for Research Use Only

Formula: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MDL Number: MFCD00054610
CAS Number: 764-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR