(4-Fluorophenyl)acetaldehyde ≥98%

Supplier: Ambeed
Warning

A450913-100MG A450913-1G A450913-25G A450913-5G
77890-913EA 22 USD
77890-913 77890-914 77890-915 77890-916
(4-Fluorophenyl)acetaldehyde ≥98%
(4-Fluorophenyl)acetaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO
MDL Number: MFCD02261761
CAS Number: 1736-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR