1-Amino-4-methylpiperazine ≥97%

Supplier: Ambeed
Warning

A450910-25G A450910-100G A450910-5G
77594-406EA 21.53 USD
77594-406 77890-911 77890-912
1-Amino-4-methylpiperazine ≥97%
1-Amino-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₅H₁₃N₃
MDL Number: MFCD00006154
CAS Number: 6928-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR