2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine ≥98%

Supplier: Ambeed
Danger

A449167-5G A449167-1G
77572-404EA 674.65 USD
77572-404 77673-108
2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine ≥98%
2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₄N
MDL Number: MFCD11226610
CAS Number: 1159512-39-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR