Imidazo[1,2-a]pyridin-5-ylmethanamine hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A449098-250MG
A449098-1G
77572-402EA
292
USD
77572-402
77616-940
Imidazo[1,2-a]pyridin-5-ylmethanamine hydrochloride ≥95%
Imidazo[1,2-a]pyridin-5-ylmethanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀ClN₃ |
MDL Number:
MFCD30189158 CAS Number: 2231153-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...