(S)-7-(Hydroxymethyl)-2-(2-hydroxypropan-2-yl)-4-methoxy-2,3-dihydro-5H-furo[3,2-g]chromen-5-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Cimifugin

A440593-10MG A440593-1MG A440593-5MG
77613-356EA 57.68 USD
77613-356 77613-358 77613-360
(S)-7-(Hydroxymethyl)-2-(2-hydroxypropan-2-yl)-4-methoxy-2,3-dihydro-5H-furo[3,2-g]chromen-5-one ≥98%
(S)-7-(Hydroxymethyl)-2-(2-hydroxypropan-2-yl)-4-methoxy-2,3-dihydro-5H-furo[3,2-g]chromen-5-one

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₆
MW: 306.31 g/mol
MDL Number: MFCD08460265
CAS Number: 37921-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR