3-(Aminomethyl)-5-fluoropyridine ≥98%

Supplier: Ambeed
Danger

A448620-1G A448620-250MG A448620-100MG A448620-5G
77613-382EA 138.02 USD
77613-382 77613-384 77673-106 77882-162
3-(Aminomethyl)-5-fluoropyridine ≥98%
3-(Aminomethyl)-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD08450424
CAS Number: 23586-96-1
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR