tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (pyrrolidin-3-ylmethyl)carbamate hydrochloride

A440246-5G A440246-1G A440246-250MG
77613-354EA 401.7 USD
77613-354 77673-082 77673-084
tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride ≥98%
tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂·ClH
MDL Number: MFCD11101394
CAS Number: 1075260-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR