Ethyl 3-ethyl-1H-pyrazole-4-carboxylate ≥97%
Supplier: Ambeed
|
Warning
|
A448116-5G
A448116-1G
A448116-250MG
77669-010EA
314
USD
77669-010
77879-981
77879-982
Ethyl 3-ethyl-1H-pyrazole-4-carboxylate ≥97%
Ethyl 3-ethyl-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O₂ |
MDL Number:
MFCD07367317 CAS Number: 73981-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...