1-Boc-7-chloroindole-3-boronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

A447964-1G A447964-250MG A447964-5G
77884-149EA 207 USD
77884-149 77884-150 77884-151
1-Boc-7-chloroindole-3-boronic acid pinacol ester ≥97%
1-Boc-7-chloroindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₅BClNO₄
MDL Number: MFCD15143613
CAS Number: 1218790-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR