8-(Isoindolin-2-ylmethyl)-1,2-dihydro-3H-pyridazino[3,4,5-de]quinazolin-3-one ≥98%

Supplier: Ambeed
Danger

Synonyms: Stenoparib

A439847-2MG A439847-10MG A439847-5MG
77746-899EA 41 USD
77746-899 77746-898 77746-900
8-(Isoindolin-2-ylmethyl)-1,2-dihydro-3H-pyridazino[3,4,5-de]quinazolin-3-one ≥98%
8-(Isoindolin-2-ylmethyl)-1,2-dihydro-3H-pyridazino[3,4,5-de]quinazolin-3-one

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₅O
MW: 317.34 g/mol
MDL Number: MFCD28167842
CAS Number: 1140964-99-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR