(2-Dicyclohexylphosphinobiphenyl)gold(I) bis(trifluoromethanesulfonyl)imide ≥98%

Supplier: Ambeed
Warning

A439794-1G A439794-100MG A439794-250MG
77748-895EA 595 USD
77748-895 77748-894 77748-896
(2-Dicyclohexylphosphinobiphenyl)gold(I) bis(trifluoromethanesulfonyl)imide ≥98%
(2-Dicyclohexylphosphinobiphenyl)gold(I) bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₂₆H₃₁AuF₆NO₄PS₂
MDL Number: MFCD22124441
CAS Number: 1016161-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR