N-Formylglycine ethyl ester ≥98%

Supplier: Ambeed
Warning

Synonyms: N-FormylglycineEthylEster, Formamidoacetic Acid Ethyl Ester, For-Gly-OEt, Ethyl Formamidoacetate

A439789-5G A439789-100G A439789-25G
77601-912EA 12.3 USD
77601-912 77668-972 77668-974
N-Formylglycine ethyl ester ≥98%
N-Formylglycine ethyl ester

Precaución: for Research Use Only

Formula: C₅H₉NO₃
MW: 131.13 g/mol
MDL Number: MFCD00014125
CAS Number: 3154-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR